missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrobenzyl Chloride 98.0+%, TCI America™
Supplier: TCI America C0229250G
Specifications
4-Nitrobenzyl Chloride | |
73°C | |
C7H6ClNO2 | |
250 g | |
4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x | |
C1=CC(=CC=C1CCl)[N+](=O)[O-] | |
171.58 | |
CHEBI:87406 | |
≥98.0% (GC) |
100-14-1 | |
White-Yellow | |
MFCD00007374 | |
3261 | |
KGCNHWXDPDPSBV-UHFFFAOYSA-N | |
1-(chloromethyl)-4-nitrobenzene | |
7482 | |
171.58 | |
Crystalline Powder |
Chemical Identifiers
100-14-1 | |
171.58 | |
KGCNHWXDPDPSBV-UHFFFAOYSA-N | |
7482 | |
1-(chloromethyl)-4-nitrobenzene |
C7H6ClNO2 | |
MFCD00007374 | |
4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x | |
CHEBI:87406 | |
C1=CC(=CC=C1CCl)[N+](=O)[O-] |
Safety and Handling
EINECSNumber : (3)-0449
RTECSNumber : XS9093000
TSCA : Yes