Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrobenzyl bromoacetate, 98+%

$46.26 - $473.12

Chemical Identifiers

Chemical Name or Material 4-Nitrobenzyl bromoacetate
CAS 16869-24-2
Molecular Formula C9H8BrNO4
Formula Weight 274.08
MDL Number MFCD00051838
Beilstein 2531330
Synonym Bromoacetic acid 4-nitrobenzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0805804

  SDS
alfa aesar™
L0805804
2g Each for $46.26

AAL0805809

  SDS
alfa aesar™
L0805809
10g Each for $132.28

AAL0805818

  SDS
alfa aesar™
L0805818
50g Each for $473.12
Description & Specifications

Specifications

Formula Weight 274.08
Chemical Name or Material 4-Nitrobenzyl bromoacetate
Assay ≥98%
Molecular Formula C9H8BrNO4
MDL Number MFCD00051838
CAS 16869-24-2
Synonym Bromoacetic acid 4-nitrobenzyl ester
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Beilstein 2531330
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 105° to 106°C (5mmHg)
Melting Point 48° to 52°C
Quantity 2g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.