Please login to your online account to display your discounted pricing

4-Nitrobenzyl bromide, 99%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  128531000

 View more versions of this product

Catalog No. AC128531000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 216.03
Physical Form Crystalline Powder
Assay ≥98.5%
Quantity 100g
Color Light yellow to beige
Melting Point 98° to 100°C
Linear Formula O2NC6H4CH2Br
Chemical Name or Material 4-Nitrobenzyl bromide
CAS 100-11-8
Synonym α-Bromo-p-nitrotoluene
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H6BrNO2
Health Hazard 1 Danger

Specifications

Chemical Name or Material 4-Nitrobenzyl bromide
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6BrNO2
Linear Formula O2NC6H4CH2Br
Formula Weight 216.03
Synonym α-Bromo-p-nitrotoluene
Accessories

Danger

SDS & Safety Documents