Learn More
4-Nitrobenzyl bromide, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1523614
Description
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5â-uronamides as selective A3 adenosine agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5â-uronamides as selective A3 adenosine agonists.
Solubility
It hydrolyzes in water. Soluble in alcohol and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
Specifications
4-Nitrobenzyl bromide | |
97°C to 101°C | |
MFCD00007373 | |
UN3261 | |
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide | |
VOLRSQPSJGXRNJ-UHFFFAOYSA-N | |
1-(bromomethyl)-4-nitrobenzene | |
66011 | |
216.04 |
100-11-8 | |
C7H6BrNO2 | |
25 g | |
742796 | |
It hydrolyzes in water. Soluble in alcohol and ether. | |
[O-][N+](=O)C1=CC=C(CBr)C=C1 | |
216.03 | |
CHEBI:62822 | |
≥97% |
Chemical Identifiers
100-11-8 | |
216.03 | |
VOLRSQPSJGXRNJ-UHFFFAOYSA-N | |
66011 | |
1-(bromomethyl)-4-nitrobenzene |
C7H6BrNO2 | |
MFCD00007373 | |
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide | |
CHEBI:62822 | |
[O-][N+](=O)C1=CC=C(CBr)C=C1 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H318
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 202-820-6
RTECSNumber : XS7967000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only