missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrobenzyl bromide, 97+%

Manufacturer:  Alfa Aesar™ A1523636

 View more versions of this product

Catalog No. AAA1523636



Description

Description

Applications
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

Solubility
It hydrolyzes in water. Soluble in alcohol and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
Specifications

Specifications

4-Nitrobenzyl bromide
500g
C7H6BrNO2
UN3261
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
C1=CC(=CC=C1CBr)[N+](=O)[O-]
216.034
CHEBI:62822
≥97%
97°C to 101°C
100-11-8
MFCD00007373
742796
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
1-(bromomethyl)-4-nitrobenzene
66011
216.04

Chemical Identifiers

100-11-8
216.034
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
66011
1-(bromomethyl)-4-nitrobenzene
C7H6BrNO2
MFCD00007373
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
CHEBI:62822
C1=CC(=CC=C1CBr)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : XS7967000

TSCA : TSCA

Documents