Learn More
4-Nitrobenzyl alcohol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1574236
Description
4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.
Solubility
Soluble in water (2 mg/ml at 20°C).
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
4-Nitrobenzyl alcohol | |
92°C to 95°C | |
180°C (356°F) | |
MFCD00007376 | |
1424026 | |
Soluble in water (2mg/ml at 20°C). | |
OCC1=CC=C(C=C1)[N+]([O-])=O | |
153.14 | |
CHEBI:41214 | |
99% |
619-73-8 | |
185°C (12 mmHg) | |
C7H7NO3 | |
500 g | |
4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m | |
JKTYGPATCNUWKN-UHFFFAOYSA-N | |
(4-nitrophenyl)methanol | |
69275 | |
153.14 |
Chemical Identifiers
619-73-8 | |
153.14 | |
JKTYGPATCNUWKN-UHFFFAOYSA-N | |
69275 | |
(4-nitrophenyl)methanol |
C7H7NO3 | |
MFCD00007376 | |
4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m | |
CHEBI:41214 | |
OCC1=CC=C(C=C1)[N+]([O-])=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 210-611-6
RTECSNumber : DP0657100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only