missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrobenzoylglycylglycine, TCI America™

$94.72 - $802.09

Chemical Identifiers

CAS 78196-53-9
Molecular Formula C11H11N3O6
Molecular Weight (g/mol) 281.224
MDL Number MFCD00059183
InChI Key WQJHGFSTRONKLC-UHFFFAOYSA-N
PubChem CID 591728
IUPAC Name 2-[[2-[(4-nitrobenzoyl)amino]acetyl]amino]acetic acid
SMILES C1=CC(=CC=C1C(=O)NCC(=O)NCC(=O)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N017725G View Documents TCI America
N017725G
25g Each for $802.09
 
N01771G View Documents TCI America
N01771G
1g Each for $94.72
 
Specifications

Chemical Identifiers

78196-53-9
281.224
WQJHGFSTRONKLC-UHFFFAOYSA-N
2-[[2-[(4-nitrobenzoyl)amino]acetyl]amino]acetic acid
C11H11N3O6
MFCD00059183
591728
C1=CC(=CC=C1C(=O)NCC(=O)NCC(=O)O)[N+](=O)[O-]

Specifications

White-Yellow
25g
C11H11N3O6
WQJHGFSTRONKLC-UHFFFAOYSA-N
2-[[2-[(4-nitrobenzoyl)amino]acetyl]amino]acetic acid
591728
Crystal-Powder at 20°C
4-Nitrobenzoylglycylglycine
218°C
78196-53-9
MFCD00059183
C1=CC(=CC=C1C(=O)NCC(=O)NCC(=O)O)[N+](=O)[O-]
281.224
281.22
≥99.0% (T)
Safety and Handling

Safety and Handling

TSCA : No