Learn More
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150430050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Nitrobenzoyl chloride | |
122-04-3 | |
100.0 | |
Yellow | |
102°C | |
98% | |
C7H4ClNO3 | |
MFCD00007345 | |
09, 394 | |
15, 6677 | |
Solubility in water: decomposes in water. Other solubilities: decomposes in alcohol,soluble in diethyl ether | |
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O | |
185.56 | |
185.57 | |
Crystals or Flakes |
98% | |
97.5 | |
70°C to 74°C | |
202°C to 205°C (105.0 mmHg) | |
Authentic | |
Glass bottle | |
O2NC6H4COCl | |
5 g | |
01,736 | |
p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride | |
SKDHHIUENRGTHK-UHFFFAOYSA-N | |
4-nitrobenzoyl chloride | |
8502 | |
98% |
Chemical Identifiers
122-04-3 | |
185.56 | |
SKDHHIUENRGTHK-UHFFFAOYSA-N | |
8502 | |
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O |
C7H4ClNO3 | |
MFCD00007345 | |
p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride | |
4-nitrobenzoyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 204-517-4
RTECSNumber : DM6651000
TSCA : TSCA
RUO – Research Use Only