Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrobenzonitrile, 97%

$73.69 - $73.69

Chemical Identifiers

Chemical Name or Material 4-Nitrobenzonitrile
CAS 619-72-7
Molecular Formula C7H4N2O2
Formula Weight 148.12
MDL Number MFCD00007279
UN Number UN3439
Beilstein 972078
Synonym 4-Cyanonitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1278314

  SDS
alfa aesar™
A1278314
25g Each for $73.69

AAA1278322

  SDS
alfa aesar™
A1278322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.12
UN Number UN3439
Health Hazard 2 GHS H Statement
H300-H311
Fatal if swallowed.
Toxic in contact with skin.
Beilstein 972078
Chemical Name or Material 4-Nitrobenzonitrile
Odor Almond-like
TSCA TSCA
Melting Point 143° to 147°C
Assay 97%
Health Hazard 1 Danger
Quantity 25g
Health Hazard 3 GHS P Statement
P280h-P309-P310-P302+P352
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
CAS 619-72-7
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
RTECS Number DI4903500
Molecular Formula C7H4N2O2
MDL Number MFCD00007279
Synonym 4-Cyanonitrobenzene
Safety and Handling

Danger

GHS P Statement
P280h-P309-P310-P302+P352
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H300-H311
Fatal if swallowed.
Toxic in contact with skin.