Please login to your online account to display your discounted pricing

4-Nitrobenzoic Acid 99+%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  128460250

 View more versions of this product

Catalog No. AC128460250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 167.12
Quantity 25g
Color Light yellow
RTECS Number DH5075000
TSCA TSCA
EINECS Number 200-526-2
Melting Point 239° to 242°C
CAS Min % 99.0
CAS Max % 100.0
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Toxic in contact with skin.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Linear Formula O2NC6H4CO2H
Name Note 99+%
MDL Number MFCD00007352
CAS 62-23-7
Chemical Name or Material 4-Nitrobenzoic acid
Molecular Formula C7H5NO4
Assay Percent Range 99+%

Specifications

Chemical Name or Material 4-Nitrobenzoic acid
Name Note 99+%
CAS 62-23-7
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H5NO4
Linear Formula O2NC6H4CO2H
Formula Weight 167.12
MDL Number MFCD00007352
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Toxic in contact with skin.
May cause respiratory irritation.

SDS & Safety Documents