Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrobenzo-15-crown-5, 99%

$61.32 - $268.20

Chemical Identifiers

Chemical Name or Material 4-Nitrobenzo-15-crown-5
CAS 60835-69-0
Molecular Formula C14H19NO7
Formula Weight 313.3
MDL Number MFCD00060713
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1622503

 
alfa aesar™
A1622503
1g Each for $61.32

AAA1622506

 
alfa aesar™
A1622506
5g Each for $268.20
Description & Specifications

Specifications

Formula Weight 313.3
Assay 99%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C14H19NO7
Melting Point 93° to 95°C
MDL Number MFCD00060713
CAS 60835-69-0
Chemical Name or Material 4-Nitrobenzo-15-crown-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.

Solubility
It is soluble in non-polar solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.