Please login to your online account to display your discounted pricing

4-Nitrobenzenesulfonyl Fluoride 99%, ACROS Organics™

$251.12 - $251.12

Chemical Identifiers

Chemical Name or Material 4-Nitrobenzenesulfonyl fluoride
Name Note 99%
CAS 349-96-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H4FNO4S
Linear Formula SO2FC6H4NO2
Formula Weight 205.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC415860250

  SDS
thermo fisher scientific
415860250
Glass bottle 25g Each for $251.12
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 205.17
Physical Form Crystalline Powder
Quantity 25g
Color Yellow
Melting Point 76° to 80°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00009637
CAS 349-96-2
Linear Formula SO2FC6H4NO2
Name Note 99%
Chemical Name or Material 4-Nitrobenzenesulfonyl fluoride
Molecular Formula C6H4FNO4S
Assay Percent Range 99%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.