Please login to your online account to display your discounted pricing

4-Nitrobenzenesulfonyl Chloride 95%, ACROS Organics™

$58.28 - $192.86

Chemical Identifiers

Chemical Name or Material 4-Nitrobenzenesulfonyl chloride
Name Note 95%
CAS 98-74-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H4ClNO4S
Linear Formula SO2ClC6H4NO2
Formula Weight 221.62
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC415850250

  SDS
thermo fisher scientific
415850250
Glass bottle 25g Each for $58.28

AC415851000

  SDS
thermo fisher scientific
415851000
Glass bottle 100g Each for $192.86

AC415850050

  SDS
thermo fisher scientific
AC415850050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 221.62
Physical Form Powder
Quantity 25g
Color Yellow to brown
Beilstein 11, 72
EINECS Number 202-697-9
Melting Point 75° to 80°C
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 1 GHS Signal Word: Danger
Name Note 95%
Molecular Formula C6H4ClNO4S
MDL Number MFCD00007445
Linear Formula SO2ClC6H4NO2
Assay Percent Range 95%
CAS 98-74-8
Chemical Name or Material 4-Nitrobenzenesulfonyl chloride
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.