missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrobenzenesulfonamide, 97%

$69.85 - $243.65

Chemical Identifiers

CAS 6325-93-5
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.184
MDL Number MFCD00007937
InChI Key QWKKYJLAUWFPDB-UHFFFAOYSA-N
Synonym 4-nitro-benzenesulfonamide
PubChem CID 22784
IUPAC Name 4-nitrobenzenesulfonamide
SMILES C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1810106 View Documents Alfa Aesar™
A1810106
5g Each for $69.85
 
AAA1810114 View Documents Alfa Aesar™
A1810114
25g Each for $243.65
 
Specifications

Chemical Identifiers

6325-93-5
202.184
QWKKYJLAUWFPDB-UHFFFAOYSA-N
22784
C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)N
C6H6N2O4S
MFCD00007937
4-nitro-benzenesulfonamide
4-nitrobenzenesulfonamide

Specifications

178°C to 181°C
6325-93-5
MFCD00007937
QWKKYJLAUWFPDB-UHFFFAOYSA-N
4-nitrobenzenesulfonamide
22784
97%
5g
C6H6N2O4S
4-nitro-benzenesulfonamide
C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)N
202.184
202.19
4-Nitrobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DB2975000

TSCA : TSCA