Learn More
4-Nitrobenzenediazonium tetrafluoroborate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5531514
Description
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4'-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4′-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.
Solubility
Soluble in acetonitrile and dimethyl sulfoxide.
Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.
Specifications
4-Nitrobenzenediazonium tetrafluoroborate | |
144°C to 148°C | |
MFCD00012005 | |
UN2923 | |
4-nitrobenzenediazonium tetrafluoroborate, ccris 3641, unii-2uxo615r4o, p-nitrobenzenediazonium tetrafluoroborate, benzenediazonium, 4-nitro-, tetrafluoroborate 1-, p-nitrophenyldiazonium fluoborate, p-nitrobenzenediazonium fluoborate, 4-nitrobenzenediazonium fluoborate, p-nitrobenzenediazonium fluoroborate, 4-nitrobenzenediazonium fluoroborate | |
AFULQCYCQAHYIP-UHFFFAOYSA-N | |
4-nitrobenzenediazonium;tetrafluoroborate | |
68006 | |
97% |
456-27-9 | |
C6H4BF4N3O2 | |
25 g | |
3599869 | |
Soluble in acetonitrile and dimethyl sulfoxide. | |
F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N | |
236.92 | |
236.92 |
Chemical Identifiers
456-27-9 | |
236.92 | |
AFULQCYCQAHYIP-UHFFFAOYSA-N | |
68006 | |
F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N |
C6H4BF4N3O2 | |
MFCD00012005 | |
4-nitrobenzenediazonium tetrafluoroborate, ccris 3641, unii-2uxo615r4o, p-nitrobenzenediazonium tetrafluoroborate, benzenediazonium, 4-nitro-, tetrafluoroborate 1-, p-nitrophenyldiazonium fluoborate, p-nitrobenzenediazonium fluoborate, 4-nitrobenzenediazonium fluoborate, p-nitrobenzenediazonium fluoroborate, 4-nitrobenzenediazonium fluoroborate | |
4-nitrobenzenediazonium;tetrafluoroborate |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 207-261-1
RTECSNumber : CZ1723000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only