missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrobenzenediazonium tetrafluoroborate, 97%

Manufacturer:  Alfa Aesar™ H5531506

 View more versions of this product

Catalog No. AAH5531506



Description

Description

Applications
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4′-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

Solubility
Soluble in acetonitrile and dimethyl sulfoxide.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.
Specifications

Specifications

4-Nitrobenzenediazonium tetrafluoroborate
5g
C6H4BF4N3O2
UN2923
4-nitrobenzenediazonium fluoborate
AFULQCYCQAHYIP-UHFFFAOYSA-N
4-nitrobenzenediazonium;tetrafluoroborate
68006
97%
144°C to 148°C
456-27-9
MFCD00012005
3599869
Soluble in acetonitrile and dimethyl sulfoxide.
[B-](F)(F)(F)F.C1=CC(=CC=C1[N+]#N)[N+](=O)[O-]
236.921
236.92

Chemical Identifiers

456-27-9
236.921
AFULQCYCQAHYIP-UHFFFAOYSA-N
68006
[B-](F)(F)(F)F.C1=CC(=CC=C1[N+]#N)[N+](=O)[O-]
C6H4BF4N3O2
MFCD00012005
4-nitrobenzenediazonium fluoborate
4-nitrobenzenediazonium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 207-261-1

RTECSNumber : CZ1723000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents