missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrobenzamidine Hydrochloride, TCI America™

$84.03 - $288.75

Chemical Identifiers

CAS 15723-90-7
Molecular Formula C7H8ClN3O2
Molecular Weight (g/mol) 201.61
MDL Number MFCD00278833
InChI Key CJXJAUKCHHAJQN-UHFFFAOYSA-N
PubChem CID 170625
IUPAC Name 4-nitrobenzenecarboximidamide;hydrochloride
SMILES C1=CC(=CC=C1C(=N)N)[N+](=O)[O-].Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09895G View Documents TCI America
N09895G
5g Each for $84.03
 
N098925G View Documents TCI America
N098925G
25g Each for $288.75
 
Specifications

Chemical Identifiers

15723-90-7
201.61
CJXJAUKCHHAJQN-UHFFFAOYSA-N
4-nitrobenzenecarboximidamide;hydrochloride
C7H8ClN3O2
MFCD00278833
170625
C1=CC(=CC=C1C(=N)N)[N+](=O)[O-].Cl

Specifications

White-Yellow
5g
C7H8ClN3O2
CJXJAUKCHHAJQN-UHFFFAOYSA-N
4-nitrobenzenecarboximidamide;hydrochloride
170625
Crystal-Powder at 20°C
4-Nitrobenzamidine Hydrochloride
286°C
15723-90-7
MFCD00278833
C1=CC(=CC=C1C(=N)N)[N+](=O)[O-].Cl
201.61
201.61
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

RTECSNumber : CV6296000

TSCA : No

Recommended Storage : Freezer