missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrobenzamide, TCI America™

$111.53 - $1,029.75

Chemical Identifiers

CAS 619-80-7
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007994
InChI Key ZESWUEBPRPGMTP-UHFFFAOYSA-N
Synonym p-nitrobenzamide, benzamide, 4-nitro, benzamide, p-nitro, 4-nitro-benzamide, 4-nitro benzamide, 4-carbamoylnitrobenzene, pubchem13419, acmc-209myf, p-nitrobenzamide, 4-nitrobenzamide
PubChem CID 12091
IUPAC Name 4-nitrobenzamide
SMILES C1=CC(=CC=C1C(=O)N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0323500G View Documents TCI America
N0323500G
500g Each for $1,029.75
 
N032325G View Documents TCI America
N032325G
25g Each for $111.53
 
Specifications

Chemical Identifiers

619-80-7
166.136
ZESWUEBPRPGMTP-UHFFFAOYSA-N
12091
C1=CC(=CC=C1C(=O)N)[N+](=O)[O-]
C7H6N2O3
MFCD00007994
p-nitrobenzamide, benzamide, 4-nitro, benzamide, p-nitro, 4-nitro-benzamide, 4-nitro benzamide, 4-carbamoylnitrobenzene, pubchem13419, acmc-209myf, p-nitrobenzamide, 4-nitrobenzamide
4-nitrobenzamide

Specifications

Yellow
500g
C7H6N2O3
p-nitrobenzamide, benzamide, 4-nitro, benzamide, p-nitro, 4-nitro-benzamide, 4-nitro benzamide, 4-carbamoylnitrobenzene, pubchem13419, acmc-209myf, p-nitrobenzamide, 4-nitrobenzamide
C1=CC(=CC=C1C(=O)N)[N+](=O)[O-]
166.136
166.14
≥98.0% (N)
203°C
619-80-7
MFCD00007994
ZESWUEBPRPGMTP-UHFFFAOYSA-N
4-nitrobenzamide
12091
Crystal-Powder at 20°C
4-Nitrobenzamide
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1508

RTECSNumber : CV5601000

TSCA : Yes