Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrobenzamide, 98+%

$70.51 - $70.51

Chemical Identifiers

Chemical Name or Material 4-Nitrobenzamide
CAS 619-80-7
Molecular Formula C7H6N2O3
Formula Weight 166.14
MDL Number MFCD00007994
Beilstein 639263
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1153614

  SDS
alfa aesar™
A1153614
25g Each for $70.51
Description & Specifications

Specifications

Formula Weight 166.14
Chemical Name or Material 4-Nitrobenzamide
TSCA TSCA
Assay ≥98%
Molecular Formula C7H6N2O3
MDL Number MFCD00007994
CAS 619-80-7
Beilstein 639263
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CV5601000
Melting Point 198° to 202°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.