missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrobenzaldehyde, 99%

Manufacturer:  Alfa Aesar™ A1165514

 View more versions of this product

Catalog No. AAA1165514



Description

Description

Applications
4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.

Solubility
Soluble in water, ethanol, benzene, glacial acetic acid. Slightly soluble in ether.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications

Specifications

4-Nitrobenzaldehyde
99%
MFCD00007346
14,6587
C1=CC(=CC=C1C=O)[N+](=O)[O-]
151.121
CHEBI:66926
555-16-8
C7H5NO3
386796
BXRFQSNOROATLV-UHFFFAOYSA-N
4-nitrobenzaldehyde
541
151.12

Chemical Identifiers

555-16-8
151.121
BXRFQSNOROATLV-UHFFFAOYSA-N
CHEBI:66926
C1=CC(=CC=C1C=O)[N+](=O)[O-]
C7H5NO3
MFCD00007346
541
4-nitrobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash contaminated clothing before reuse.
Specific treatment (see ... on this label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CU7350000

TSCA : TSCA

SDS Documents