missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitroaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A103690I

 View more versions of this product

Catalog No. AAA103690I


Description

Description

p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

Solubility
Soluble in hot water and alcohol

Notes
Store at 4°C. Incompatible with oxidizing agents.
Specifications

Specifications

4-Nitroaniline
147°C to 149°C
260°C (100 mmHg)
Amine-like
MFCD00007858
UN1661
14,6584
Soluble in hot water and alcohol
NC1=CC=C(C=C1)[N+]([O-])=O
138.13
CHEBI:17064
98%
100-01-6
1.437
165°C (329°F)
C6H6N2O2
5000 g
508690
p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p
TYMLOMAKGOJONV-UHFFFAOYSA-N
4-nitroaniline
7475
138.13

Chemical Identifiers

100-01-6
138.13
TYMLOMAKGOJONV-UHFFFAOYSA-N
7475
4-nitroaniline
C6H6N2O2
MFCD00007858
p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p
CHEBI:17064
NC1=CC=C(C=C1)[N+]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROANILINES

EINECSNumber : 202-810-1

RTECSNumber : BY7000000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only