Please login to your online account to display your discounted pricing

4'-Nitroacetanilide, 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC415755000

 View more versions of this product

Catalog No. AC415755000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Solid
Formula Weight 180.16
Quantity 500g
Color Yellow to green-yellow or green-brown
Melting Point 213° to 216°C
Merck Index 15, 6667
CAS Max % 100.0
CAS 104-04-1
Chemical Name or Material 4';-Nitroacetanilide
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula CH3C(O)NHC6H4NO2
MDL Number MFCD00007303
CAS Min % 98.5
Health Hazard 1 Warning
Molecular Formula C8H8N2O3
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4';-Nitroacetanilide
CAS 104-04-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H8N2O3
Linear Formula CH3C(O)NHC6H4NO2
Formula Weight 180.16
MDL Number MFCD00007303
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents