missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitro-o-cresol, TCI America™

$71.81 - $212.36

Chemical Identifiers

CAS 99-53-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD01734013
InChI Key KDQPMQNHVQVVMR-UHFFFAOYSA-N
Synonym 2-Hydroxy-5-nitrotoluene, 2-Methyl-4-nitrophenol
PubChem CID 7442
IUPAC Name 2-methyl-4-nitrophenol
SMILES CC1=C(C=CC(=C1)[N+](=O)[O-])O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06705G View Documents TCI America
N06705G
5g Each for $71.81
 
N067025G View Documents TCI America
N067025G
25g Each for $212.36
 
Specifications

Chemical Identifiers

99-53-6
153.137
KDQPMQNHVQVVMR-UHFFFAOYSA-N
7442
CC1=C(C=CC(=C1)[N+](=O)[O-])O
C7H7NO3
MFCD01734013
2-Hydroxy-5-nitrotoluene, 2-Methyl-4-nitrophenol
2-methyl-4-nitrophenol

Specifications

Yellow
5g
C7H7NO3
2446
KDQPMQNHVQVVMR-UHFFFAOYSA-N
2-methyl-4-nitrophenol
7442
Crystal-Powder at 20°C
4-Nitro-o-cresol
96°C
99-53-6
MFCD01734013
2-Hydroxy-5-nitrotoluene, 2-Methyl-4-nitrophenol
CC1=C(C=CC(=C1)[N+](=O)[O-])O
153.137
153.14
≥98.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0790

RTECSNumber : GP2700000

TSCA : No