missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-N-n-propylbenzamide, 97%

$223.26 - $710.82

Chemical Identifiers

CAS 2585-24-2
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
MDL Number MFCD00024716
InChI Key MDRVSZJDSUDYGP-UHFFFAOYSA-N
Synonym 4-nitro-n-n-propylbenzamide, 4-nitro-n-propyl-benzamide, 4-nitrophenyl-n-propylcarboxamide, acmc-1clws, benzamide, 4-nitro-n-propyl, benzamide, p-nitro-n-propyl, benzamide,4-nitro-n-propyl, n-n-propyl4-nitrobenzamide, n-propyl-4-nitrobenzamide
PubChem CID 314664
IUPAC Name 4-nitro-N-propylbenzamide
SMILES CCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60041MD
View Documents
Alfa Aesar™
H60041MD
250mg Each for $223.26
 
AAH6004103
View Documents
Alfa Aesar™
H6004103
1g Each for $710.82
 
Specifications

Chemical Identifiers

2585-24-2
208.217
MDRVSZJDSUDYGP-UHFFFAOYSA-N
314664
CCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C10H12N2O3
MFCD00024716
4-nitro-n-n-propylbenzamide, 4-nitro-n-propyl-benzamide, 4-nitrophenyl-n-propylcarboxamide, acmc-1clws, benzamide, 4-nitro-n-propyl, benzamide, p-nitro-n-propyl, benzamide,4-nitro-n-propyl, n-n-propyl4-nitrobenzamide, n-propyl-4-nitrobenzamide
4-nitro-N-propylbenzamide

Specifications

100°C to 103°C (decomposition)
2585-24-2
MFCD00024716
MDRVSZJDSUDYGP-UHFFFAOYSA-N
4-nitro-N-propylbenzamide
314664
97%
250mg
C10H12N2O3
4-nitro-n-n-propylbenzamide, 4-nitro-n-propyl-benzamide, 4-nitrophenyl-n-propylcarboxamide, acmc-1clws, benzamide, 4-nitro-n-propyl, benzamide, p-nitro-n-propyl, benzamide,4-nitro-n-propyl, n-n-propyl4-nitrobenzamide, n-propyl-4-nitrobenzamide
CCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
208.217
208.21
4-Nitro-N-n-propylbenzamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures