missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-N-n-propylbenzamide, 97%

Manufacturer:  Alfa Aesar™ H60041MD

 View more versions of this product

Catalog No. AAH60041MD



Specifications

Specifications

4-Nitro-N-n-propylbenzamide
250mg
C10H12N2O3
4-nitro-n-n-propylbenzamide, 4-nitro-n-propyl-benzamide, 4-nitrophenyl-n-propylcarboxamide, acmc-1clws, benzamide, 4-nitro-n-propyl, benzamide, p-nitro-n-propyl, benzamide,4-nitro-n-propyl, n-n-propyl4-nitrobenzamide, n-propyl-4-nitrobenzamide
CCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
208.217
208.21
100°C to 103°C (decomposition)
2585-24-2
MFCD00024716
MDRVSZJDSUDYGP-UHFFFAOYSA-N
4-nitro-N-propylbenzamide
314664
97%

Chemical Identifiers

2585-24-2
208.217
MDRVSZJDSUDYGP-UHFFFAOYSA-N
314664
CCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C10H12N2O3
MFCD00024716
4-nitro-n-n-propylbenzamide, 4-nitro-n-propyl-benzamide, 4-nitrophenyl-n-propylcarboxamide, acmc-1clws, benzamide, 4-nitro-n-propyl, benzamide, p-nitro-n-propyl, benzamide,4-nitro-n-propyl, n-n-propyl4-nitrobenzamide, n-propyl-4-nitrobenzamide
4-nitro-N-propylbenzamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents