Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-N-(n-propyl)benzylamine hydrochloride, 99%

$49.73 - $658.46

Chemical Identifiers

Chemical Name or Material 4-Nitro-N-(n-propyl)benzylamine hydrochloride
CAS 68133-98-2
Molecular Formula C10H14N2O2·HCl
Formula Weight 230.7
MDL Number MFCD00012845
Beilstein 3714783
Synonym 4-Nitrobenzyl-n-propylamine hydrochloride; N-(n-Propyl)-4-nitrobenzylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1930803

  SDS
alfa aesar™
A1930803
1g Each for $49.73

AAA1930806

  SDS
alfa aesar™
A1930806
5g Each for $159.72

AAA1930814

  SDS
alfa aesar™
A1930814
25g Each for $658.46
Description & Specifications

Specifications

Formula Weight 230.7
Chemical Name or Material 4-Nitro-N-(n-propyl)benzylamine hydrochloride
TSCA TSCA
Assay 99%
Molecular Formula C10H14N2O2·HCl
MDL Number MFCD00012845
CAS 68133-98-2
Beilstein 3714783
Synonym 4-Nitrobenzyl-n-propylamine hydrochloride; N-(n-Propyl)-4-nitrobenzylamine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 228° to 234°C
Quantity 1g

Applications
4-Nitro-N-propylbenzylamine hydrochloride was used as a derivatization reagent for the extraction of amine in the determination of trace amounts of diisocyanate monomers in contaminated air by an electroanalytical method.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.