Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-m-xylene, 99%

Chemical Identifiers

Chemical Name or Material 4-Nitro-m-xylene
CAS 89-87-2
Molecular Formula C8H9NO2
Formula Weight 151.17g/mol
MDL Number MFCD00007169
UN Number UN1665
Beilstein 1865656
Synonym 1,3-Dimethyl-4-nitrobenzene; 2,4-Dimethylnitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1677618

 
alfa aesar™
A16776-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1677622

 
alfa aesar™
A16776-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1677630

 
alfa aesar™
A16776-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 5° to 7°C
Boiling Point 243° to 245°C
Quantity 50g
Flash Point 107°C (224°F)
Chemical Name or Material 4-Nitro-m-xylene
CAS 89-87-2
Assay 99%
Molecular Formula C8H9NO2
Formula Weight 151.17g/mol
MDL Number MFCD00007169
UN Number UN1665
Beilstein 1865656
Synonym 1,3-Dimethyl-4-nitrobenzene; 2,4-Dimethylnitrobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H331-H302
Toxic in contact with skin.
Toxic if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P261-P280-P361-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROXYLENES, LIQUID
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P361-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H331-H302
Toxic in contact with skin.
Toxic if inhaled.
Harmful if swallowed.