Learn More
4-Nitro-m-phenylenediamine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1216609
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Nitro-m-phenylenediamine | |
156°C to 160°C | |
MFCD00025289 | |
2094904 | |
DPIZKMGPXNXSGL-UHFFFAOYSA-N | |
4-nitrobenzene-1,3-diamine | |
21208 | |
95% |
5131-58-8 | |
C6H7N3O2 | |
10 g | |
4-nitro-m-phenylenediamine, 4-nitro-1,3-phenylenediamine, 2,4-diaminonitrobenzene, 1,3-benzenediamine, 4-nitro, 4-nitro-1,3-benzenediamine, 1,3-diamino-4-nitrobenzene, m-phenylenediamine, 4-nitro, 3-amino-4-nitrophenylamine, 4-nitro-1,3-fenylendiamin, unii-0d5u5ew62z | |
NC1=CC=C(C(N)=C1)[N+]([O-])=O | |
153.14 | |
153.14 |
Chemical Identifiers
5131-58-8 | |
153.14 | |
DPIZKMGPXNXSGL-UHFFFAOYSA-N | |
21208 | |
NC1=CC=C(C(N)=C1)[N+]([O-])=O |
C6H7N3O2 | |
MFCD00025289 | |
4-nitro-m-phenylenediamine, 4-nitro-1,3-phenylenediamine, 2,4-diaminonitrobenzene, 1,3-benzenediamine, 4-nitro, 4-nitro-1,3-benzenediamine, 1,3-diamino-4-nitrobenzene, m-phenylenediamine, 4-nitro, 3-amino-4-nitrophenylamine, 4-nitro-1,3-fenylendiamin, unii-0d5u5ew62z | |
4-nitrobenzene-1,3-diamine |
Safety and Handling
GHS H Statement
H301-H341-H315-H319
Toxic if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
H302-H315-H317-H319
EINECSNumber : 225-876-3
RTECSNumber : ST2910000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only