missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-L-phenylalanine monohydrate, 98%

$54.46 - $189.99

Chemical Identifiers

CAS 207591-86-4
Molecular Formula C9H10N2O4·H2O
Formula Weight 228.21 (210.19 Anhydrous)
MDL Number MFCD00051221
Beilstein 2809673
Synonym H-Phe(4-NO2)-OH
Chemical Name or Material 4-Nitro-L-phenylalanine monohydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1270903 SDS Alfa Aesar™
L1270903
1g Each for $54.46
AAL1270906 SDS Alfa Aesar™
L1270906
5g Each for $189.99
Specifications Description & Specifications

Chemical Identifiers

207591-86-4
228.21 (210.19 Anhydrous)
2809673
4-Nitro-L-phenylalanine monohydrate
C9H10N2O4·H2O
MFCD00051221
H-Phe(4-NO2)-OH

Specifications

207591-86-4
228.21 (210.19 Anhydrous)
2809673
236° to 239°C
+6.5° (c=1 in 5N HCl)
98%
C9H10N2O4·H2O
MFCD00051221
H-Phe(4-NO2)-OH
1g
4-Nitro-L-phenylalanine monohydrate

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AY7400000