missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-L-phenylalanine monohydrate, 98%

Manufacturer:  Alfa Aesar™ L1270906

 View more versions of this product

Catalog No. AAL1270906



Specifications

Specifications

4-Nitro-L-phenylalanine monohydrate
C9H12N2O5
2809673
OLZDHJRNUIXKLU-QRPNPIFTSA-N
(2S)-2-amino-3-(4-nitrophenyl)propanoic acid;hydrate
2734751
98%
5g
207591-86-4
MFCD00051221
4-nitro-l-phenylalanine monohydrate, 4-nitro-l-phenylalanine hydrate, s-2-amino-3-4-nitrophenyl propanoic acid hydrate, h-phe 4-no2-oh.h2o, 2s-2-amino-3-4-nitrophenyl propanoic acid hydrate, 4-nitro-l-phenylalanin hydrate, phenylalanine, 4-nitro-hydrate, l-phenylalanine, 4-nitro-, monohydrate, 4-nitro-l-phenylalnine, pubchem12061
C1=CC(=CC=C1CC(C(=O)O)N)[N+](=O)[O-].O
228.204
228.21 (210.19 Anhydrous)
236°C to 239°C
+6.5° (c=1 in 5N HCl)

Chemical Identifiers

207591-86-4
228.204
OLZDHJRNUIXKLU-QRPNPIFTSA-N
2734751
C1=CC(=CC=C1CC(C(=O)O)N)[N+](=O)[O-].O
C9H12N2O5
MFCD00051221
4-nitro-l-phenylalanine monohydrate, 4-nitro-l-phenylalanine hydrate, s-2-amino-3-4-nitrophenyl propanoic acid hydrate, h-phe 4-no2-oh.h2o, 2s-2-amino-3-4-nitrophenyl propanoic acid hydrate, 4-nitro-l-phenylalanin hydrate, phenylalanine, 4-nitro-hydrate, l-phenylalanine, 4-nitro-, monohydrate, 4-nitro-l-phenylalnine, pubchem12061
(2S)-2-amino-3-(4-nitrophenyl)propanoic acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AY7400000

Documents