missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-L-phenylalanine methyl ester hydrochloride, 97%

$87.12 - $332.49

Chemical Identifiers

CAS 17193-40-7
Molecular Formula C10H12N2O4·HCl
Formula Weight 260.68
MDL Number MFCD00135754
Beilstein 5176380
Synonym H-Phe(4-NO2)-OMe·HCl
Chemical Name or Material 4-Nitro-L-phenylalanine methyl ester hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0939003 SDS Alfa Aesar™
L0939003
1g Each for $87.12
AAL0939006 SDS Alfa Aesar™
L0939006
5g Each for $332.49
Specifications Description & Specifications

Chemical Identifiers

17193-40-7
260.68
5176380
4-Nitro-L-phenylalanine methyl ester hydrochloride
C10H12N2O4·HCl
MFCD00135754
H-Phe(4-NO2)-OMe·HCl

Specifications

260.68
17193-40-7
5176380
≈232°C
4-Nitro-L-phenylalanine methyl ester hydrochloride
MFCD00135754
H-Phe(4-NO2)-OMe·HCl
1g
C10H12N2O4·HCl
97%

Safety and Handling

GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash contaminated clothing before reuse.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning