Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-L-phenylalanine, 98%

$66.15 - $168.76

Chemical Identifiers

Chemical Name or Material 4-Nitro-L-phenylalanine
CAS 949-99-5
Molecular Formula C9H10N2O4
Formula Weight 210.19
MDL Number MFCD00051221
Synonym H-Phe(4-NO2)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2706703

  SDS
alfa aesar™
H2706703
1g Each for $168.76

AAH27067MD

  SDS
alfa aesar™
H27067MD
250mg Each for $66.15
Description & Specifications

Specifications

Formula Weight 210.19
Odor Odorless
Molecular Formula C9H10N2O4
Chemical Name or Material 4-Nitro-L-phenylalanine
Quantity 1g
RTECS Number AY7400000
Synonym H-Phe(4-NO2)-OH
MDL Number MFCD00051221
Assay 98%
CAS 949-99-5