missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitro-5(2H)-isoxazolone Pyridinium Salt, TCI America™

$197.09 - $536.27

Chemical Identifiers

CAS 145640-15-9
Molecular Formula C8H7N3O4
Molecular Weight (g/mol) 209.161
InChI Key CBUGMUJYFOVCQX-UHFFFAOYSA-N
PubChem CID 15193373
IUPAC Name 4-nitro-2H-1,2-oxazol-5-one;pyridine
SMILES C1=CC=NC=C1.C1=C(C(=O)ON1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10915G View Documents TCI America
N10915G
5g Each for $536.27
 
N10911G View Documents TCI America
N10911G
1g Each for $197.09
 
Specifications

Chemical Identifiers

145640-15-9
209.161
15193373
C1=CC=NC=C1.C1=C(C(=O)ON1)[N+](=O)[O-]
C8H7N3O4
CBUGMUJYFOVCQX-UHFFFAOYSA-N
4-nitro-2H-1,2-oxazol-5-one;pyridine

Specifications

White-Yellow
5g
C8H7N3O4
C1=CC=NC=C1.C1=C(C(=O)ON1)[N+](=O)[O-]
209.161
209.16
≥93.0% (T)
131°C
145640-15-9
CBUGMUJYFOVCQX-UHFFFAOYSA-N
4-nitro-2H-1,2-oxazol-5-one;pyridine
15193373
Crystal-Powder at 20°C
4-Nitro-5(2H)-isoxazolone Pyridinium Salt
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator