missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitro-3-(trifluoromethyl)phenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC319720050

 View more versions of this product

Catalog No. AC319720050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
207.11
Yellow
15, 9401
O2NC6H3(CF3)OH
99%
88-30-2
98.5
Glass bottle
Crystalline Powder
5g
72° to 79°C
100.0
Authentic
C7H4F3NO3
4-Nitro-3-(trifluoromethyl)phenol
MFCD00014714

Chemical Identifiers

88-30-2
100.0
C7H4F3NO3
207.11
4-Nitro-3-(trifluoromethyl)phenol
98.5
99%
O2NC6H3(CF3)OH
MFCD00014714
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.