Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-3-picoline N-oxide, 98%

Chemical Identifiers

Chemical Name or Material 4-Nitro-3-picoline N-oxide
CAS 1074-98-2
Molecular Formula C6H6N2O3
Formula Weight 154.13
MDL Number MFCD00014626
UN Number UN2811
Beilstein 140157
Synonym 3-Methyl-4-nitropyridine 1-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0609903

  SDS
alfa aesar™
L0609903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0609906

  SDS
alfa aesar™
L0609906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.13
Chemical Name or Material 4-Nitro-3-picoline N-oxide
CAS 1074-98-2
Molecular Formula C6H6N2O3
Assay 98%
Synonym 3-Methyl-4-nitropyridine 1-oxide
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Beilstein 140157
Health Hazard 1 Danger
MDL Number MFCD00014626
Health Hazard 2 GHS H Statement
H311-H341-H302-H332
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Quantity 1g
Melting Point 134° to 138°C
RTECS Number UT5775000
UN Number UN2811
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.
Safety and Handling

Danger

GHS P Statement
P280h
Wear protective gloves/clothing.

GHS H Statement
H311-H341-H302-H332
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.