missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-2-(trifluoromethyl)benzoxazole, 97%

$78.80 - $1,040.93

Chemical Identifiers

CAS 573759-00-9
Molecular Formula C8H3F3N2O3
Molecular Weight (g/mol) 232.118
InChI Key MVYGNYLHXPFQJV-UHFFFAOYSA-N
Synonym 4-nitro-2-trifluoromethyl benzo d oxazole, 4-nitro-2-trifluoromethylbenzoxazole, 4-nitro-2-trifluoromethyl-1,3-benzoxazole, 4-nitro-2-trifluoromethyl benzoxazole, 4-nitro-2-trifluoromethyl-benzooxazole
PubChem CID 21875019
IUPAC Name 4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
SMILES C1=CC(=C2C(=C1)OC(=N2)C(F)(F)F)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63601MD View Documents Alfa Aesar™
H63601MD
250mg Each for $78.80
 
AAH6360103 View Documents Alfa Aesar™
H6360103
1g Each for $249.82
 
AAH6360106 View Documents Alfa Aesar™
H6360106
5g Each for $1,040.93
 
Specifications

Chemical Identifiers

573759-00-9
232.118
4-nitro-2-trifluoromethyl benzo d oxazole, 4-nitro-2-trifluoromethylbenzoxazole, 4-nitro-2-trifluoromethyl-1,3-benzoxazole, 4-nitro-2-trifluoromethyl benzoxazole, 4-nitro-2-trifluoromethyl-benzooxazole
4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
C8H3F3N2O3
MVYGNYLHXPFQJV-UHFFFAOYSA-N
21875019
C1=CC(=C2C(=C1)OC(=N2)C(F)(F)F)[N+](=O)[O-]

Specifications

250mg
C8H3F3N2O3
MVYGNYLHXPFQJV-UHFFFAOYSA-N
4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
21875019
Solid
4-Nitro-2-(trifluoromethyl)benzoxazole
573759-00-9
4-nitro-2-trifluoromethyl benzo d oxazole, 4-nitro-2-trifluoromethylbenzoxazole, 4-nitro-2-trifluoromethyl-1,3-benzoxazole, 4-nitro-2-trifluoromethyl benzoxazole, 4-nitro-2-trifluoromethyl-benzooxazole
C1=CC(=C2C(=C1)OC(=N2)C(F)(F)F)[N+](=O)[O-]
232.118
232.12
97%