missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-2-(trifluoromethyl)benzoxazole, 97%

Manufacturer:  Alfa Aesar™ H63601MD

 View more versions of this product

Catalog No. AAH63601MD



Specifications

Specifications

4-Nitro-2-(trifluoromethyl)benzoxazole
573759-00-9
4-nitro-2-trifluoromethyl benzo d oxazole, 4-nitro-2-trifluoromethylbenzoxazole, 4-nitro-2-trifluoromethyl-1,3-benzoxazole, 4-nitro-2-trifluoromethyl benzoxazole, 4-nitro-2-trifluoromethyl-benzooxazole
C1=CC(=C2C(=C1)OC(=N2)C(F)(F)F)[N+](=O)[O-]
232.118
232.12
97%
250mg
C8H3F3N2O3
MVYGNYLHXPFQJV-UHFFFAOYSA-N
4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
21875019
Solid

Chemical Identifiers

573759-00-9
232.118
4-nitro-2-trifluoromethyl benzo d oxazole, 4-nitro-2-trifluoromethylbenzoxazole, 4-nitro-2-trifluoromethyl-1,3-benzoxazole, 4-nitro-2-trifluoromethyl benzoxazole, 4-nitro-2-trifluoromethyl-benzooxazole
4-nitro-2-(trifluoromethyl)-1,3-benzoxazole
C8H3F3N2O3
MVYGNYLHXPFQJV-UHFFFAOYSA-N
21875019
C1=CC(=C2C(=C1)OC(=N2)C(F)(F)F)[N+](=O)[O-]
Documents