missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitro-2-(trifluoromethyl)benzonitrile, TCI America™

$192.50 - $575.98

Chemical Identifiers

CAS 320-47-8
Molecular Formula C8H3F3N2O2
Molecular Weight (g/mol) 216.119
MDL Number MFCD01075729
InChI Key AGKQJEFSEQHGTA-UHFFFAOYSA-N
Synonym 4-nitro-2-trifluoromethyl benzonitrile, 2-cyano-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitrobenzonitrile, benzonitrile, 4-nitro-2-trifluoromethyl, 4-nitro-2-trifluoromethyl benzenecarbonitrile, 2-trifluoromethyl-4-nitrobenzonitrile, pubchem4821, acmc-1ctuy, buttpark 91\04-59, 4-cyano-3-trifluoromethyl nitrobenzene
PubChem CID 2775783
IUPAC Name 4-nitro-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10685G View Documents TCI America
N10685G
5g Each for $192.50
 
N106825G View Documents TCI America
N106825G
25g Each for $575.98
 
Specifications

Chemical Identifiers

320-47-8
216.119
AGKQJEFSEQHGTA-UHFFFAOYSA-N
2775783
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)C#N
C8H3F3N2O2
MFCD01075729
4-nitro-2-trifluoromethyl benzonitrile, 2-cyano-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitrobenzonitrile, benzonitrile, 4-nitro-2-trifluoromethyl, 4-nitro-2-trifluoromethyl benzenecarbonitrile, 2-trifluoromethyl-4-nitrobenzonitrile, pubchem4821, acmc-1ctuy, buttpark 91\04-59, 4-cyano-3-trifluoromethyl nitrobenzene
4-nitro-2-(trifluoromethyl)benzonitrile

Specifications

Undesignated
5g
C8H3F3N2O2
3439
AGKQJEFSEQHGTA-UHFFFAOYSA-N
4-nitro-2-(trifluoromethyl)benzonitrile
2775783
Crystal-Powder at 20°C
4-Nitro-2-(trifluoromethyl)benzonitrile
51°C
320-47-8
MFCD01075729
4-nitro-2-trifluoromethyl benzonitrile, 2-cyano-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitrobenzonitrile, benzonitrile, 4-nitro-2-trifluoromethyl, 4-nitro-2-trifluoromethyl benzenecarbonitrile, 2-trifluoromethyl-4-nitrobenzonitrile, pubchem4821, acmc-1ctuy, buttpark 91\04-59, 4-cyano-3-trifluoromethyl nitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)C#N
216.119
216.12
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No