Learn More
4-Nitro-2-(trifluoromethyl)benzoic acid, 97%, ACROS Organics™
Chemical Identifiers
CAS | 320-37-6 |
---|---|
Molecular Formula | C8H4F3NO4 |
Molecular Weight (g/mol) | 235.12 |
MDL Number | MFCD04039246 |
InChI Key | BPCKZQCTLCTDST-UHFFFAOYSA-N |
Synonym | 2-carboxy-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitrobenzoic acid, 4-nitro-2-trifluoromethyl benzoic acid, 4-nitro-2-trifluoromethyl benzoic acid&, 4-nitro-2-trifluoromethyl-benzoic acid, 4-nitro-2-trifluoromethylbenzoic acid, benzoic acid, 4-nitro-2-trifluoromethyl, ksc496i1p, pubchem1342, rarechem al bo 1444 |
PubChem CID | 2782737 |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC399870010
|
Thermo Fisher Scientific
AC399870010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC399870050
|
Thermo Fisher Scientific
AC399870050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
320-37-6 | |
235.12 | |
BPCKZQCTLCTDST-UHFFFAOYSA-N | |
2782737 |
C8H4F3NO4 | |
MFCD04039246 | |
2-carboxy-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitrobenzoic acid, 4-nitro-2-trifluoromethyl benzoic acid, 4-nitro-2-trifluoromethyl benzoic acid&, 4-nitro-2-trifluoromethyl-benzoic acid, 4-nitro-2-trifluoromethylbenzoic acid, benzoic acid, 4-nitro-2-trifluoromethyl, ksc496i1p, pubchem1342, rarechem al bo 1444 | |
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)C(=O)O |
Specifications
Authentic | |
White | |
1g | |
96.0 | |
96% min. (GC) | |
O2NC6H3(CF3)CO2H | |
2-carboxy-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitrobenzoic acid, 4-nitro-2-trifluoromethyl benzoic acid, 4-nitro-2-trifluoromethyl benzoic acid&, 4-nitro-2-trifluoromethyl-benzoic acid, 4-nitro-2-trifluoromethylbenzoic acid, benzoic acid, 4-nitro-2-trifluoromethyl, ksc496i1p, pubchem1342, rarechem al bo 1444 | |
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)C(=O)O | |
2782737 | |
Crystalline Powder | |
4-Nitro-2-(trifluoromethyl)benzoic acid |
Glass bottle | |
138.0°C to 142.0°C | |
320-37-6 | |
100.0 | |
C8H4F3NO4 | |
MFCD04039246 | |
BPCKZQCTLCTDST-UHFFFAOYSA-N | |
235.12 | |
235.12 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning