missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-2-(trifluoromethyl)anisole, 98+%

Chemical Identifiers

CAS 654-76-2
Molecular Formula C8H6F3NO3
Formula Weight 221.14
MDL Number MFCD00007328
UN Number UN1759
Beilstein 2129987
Synonym 2-Methoxy-5-nitrobenzotrifluoride
Chemical Name or Material 4-Nitro-2-(trifluoromethyl)anisole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2150206 Alfa Aesar™
B2150206
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

654-76-2
221.14
UN1759
2-Methoxy-5-nitrobenzotrifluoride
C8H6F3NO3
MFCD00007328
2129987
4-Nitro-2-(trifluoromethyl)anisole

Specifications

654-76-2
221.14
UN1759
79° to 81°C
5g
C8H6F3NO3
MFCD00007328
2129987
2-Methoxy-5-nitrobenzotrifluoride
4-Nitro-2-(trifluoromethyl)anisole