Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Nitro-2'-(trifluoromethyl)acetanilide, 97%

$32.09 - $372.18

Chemical Identifiers

Chemical Name or Material 4'-Nitro-2'-(trifluoromethyl)acetanilide
CAS 395-68-6
Molecular Formula C9H7F3N2O3
Formula Weight 248.16
MDL Number MFCD00052846
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2750403

  SDS
alfa aesar™
H2750403
1g Each for $32.09

AAH2750406

  SDS
alfa aesar™
H2750406
5g Each for $102.01

AAH2750414

  SDS
alfa aesar™
H2750414
25g Each for $372.18
Description & Specifications

Specifications

Formula Weight 248.16
Chemical Name or Material 4'-Nitro-2'-(trifluoromethyl)acetanilide
TSCA TSCA
Assay 97%
Molecular Formula C9H7F3N2O3
MDL Number MFCD00052846
CAS 395-68-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 146° to 150°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.