Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitro-2-picoline N-oxide, 98%

$39.57 - $397.14

Chemical Identifiers

Chemical Name or Material 4-Nitro-2-picoline N-oxide
CAS 5470-66-6
Molecular Formula C6H6N2O3
Formula Weight 154.13
MDL Number MFCD00014625
Beilstein 135501
Synonym 2-Methyl-4-nitropyridine 1-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0661403

  SDS
alfa aesar™
L0661403
1g Each for $39.57

AAL0661414

  SDS
alfa aesar™
L0661414
25g Each for $397.14
Description & Specifications

Specifications

Formula Weight 154.13
Chemical Name or Material 4-Nitro-2-picoline N-oxide
CAS 5470-66-6
Health Hazard 2 GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Molecular Formula C6H6N2O3
MDL Number MFCD00014625
Beilstein 135501
Synonym 2-Methyl-4-nitropyridine 1-oxide
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UT5710000
Quantity 1g
Health Hazard 1 Warning
Melting Point 154° to 158°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.