missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitro-2-picoline N-oxide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441870250

 View more versions of this product

Catalog No. AC441870250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Nitro-2-picoline N-oxide
Authentic
155°C to 157°C
5470-66-6
100.0
C6H6N2O3
FTTIAVRPJGCXAC-UHFFFAOYSA-N
2-methyl-4-nitro-1-oxidopyridin-1-ium
95291
Crystalline Powder
44.89 to 48.63% C, 17.45 to 18.90% N
Yellow
25g
96.0
96% min. (HPLC)
2-methyl-4-nitropyridin-1-ium-1-olate, 2-methyl-4-nitropyridine 1-oxide, 2-methyl-4-nitropyridine n-oxide, 2-methyl-4-nitropyridine-n-oxide, 2-methyl-4-nitropyridinium-1-olate, 4-nitro-2-methylpyridine-n-oxide, 4-nitro-2-picoline n-oxide, 4-nitro-2-picoline-n-oxide, 4-nitropyridine-2-picoline-n-oxide, pyridine, 2-methyl-4-nitro-, 1-oxide
CC1=[N+](C=CC(=C1)[N+](=O)[O-])[O-]
154.13
154.13
97%

Chemical Identifiers

5470-66-6
154.13
2-methyl-4-nitropyridin-1-ium-1-olate, 2-methyl-4-nitropyridine 1-oxide, 2-methyl-4-nitropyridine n-oxide, 2-methyl-4-nitropyridine-n-oxide, 2-methyl-4-nitropyridinium-1-olate, 4-nitro-2-methylpyridine-n-oxide, 4-nitro-2-picoline n-oxide, 4-nitro-2-picoline-n-oxide, 4-nitropyridine-2-picoline-n-oxide, pyridine, 2-methyl-4-nitro-, 1-oxide
2-methyl-4-nitro-1-oxidopyridin-1-ium
C6H6N2O3
FTTIAVRPJGCXAC-UHFFFAOYSA-N
95291
CC1=[N+](C=CC(=C1)[N+](=O)[O-])[O-]
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap

GHS Signal Word: Warning

SDS Documents