Please login to your online account to display your discounted pricing

4-Nitro-2-picoline N-oxide, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Nitro-2-picoline N-oxide
CAS 5470-66-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H6N2O3
Formula Weight 154.13
Synonym 2-Methyl-4-nitropyridine N-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441870050

  SDS
thermo fisher scientific
AC441870050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441870250

  SDS
thermo fisher scientific
AC441870250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 154.13
Assay ≥96.0%
Quantity 5g
Color Pale yellow
Melting Point 154° to 160°C
CAS Max % 100.0
CAS Min % 96.0
Molecular Formula C6H6N2O3
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Suspected of causing genetic defects.
Health Hazard 1 GHS Signal Word: Warning
CAS 5470-66-6
Chemical Name or Material 4-Nitro-2-picoline N-oxide
Assay Percent Range 97%
Elemental Analysis 44.89 to 48.63% C, 17.45 to 18.90% N
Synonym 2-Methyl-4-nitropyridine N-oxide
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Suspected of causing genetic defects.