missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitro-2,3-lutidine N-oxide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441980250

 View more versions of this product

Catalog No. AC441980250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Nitro-2, 3-lutidine N-oxide
Yellow
25g
96.0
96% min. (HPLC)
2,3-dimethyl-4-nitro-1-oxido-pyridin-1-ium, 2,3-dimethyl-4-nitro-pyridine 1-oxide, 2,3-dimethyl-4-nitropyridin-1-ium-1-olate, 2,3-dimethyl-4-nitropyridine 1-oxide, 2,3-dimethyl-4-nitropyridine n-oxide, 2,3-dimethyl-4-nitropyridine-n-oxide, 4-nitro-2,3-dimethylpyridine n-oxide, 4-nitro-2,3-lutidine n-oxide, 4-nitro-2,3-lutidine-n-oxide, pyridine, 2,3-dimethyl-4-nitro-, 1-oxide
CC1=C(C=C[N+](=C1C)[O-])[N+](=O)[O-]
168.15
168.15
97%
Authentic
94°C to 96°C
37699-43-7
100.0
C7H8N2O3
CFMTVTYBZMKULI-UHFFFAOYSA-N
2,3-dimethyl-4-nitro-1-oxidopyridin-1-ium
148223
Powder or Crystalline Powder

Chemical Identifiers

37699-43-7
168.15
2,3-dimethyl-4-nitro-1-oxido-pyridin-1-ium, 2,3-dimethyl-4-nitro-pyridine 1-oxide, 2,3-dimethyl-4-nitropyridin-1-ium-1-olate, 2,3-dimethyl-4-nitropyridine 1-oxide, 2,3-dimethyl-4-nitropyridine n-oxide, 2,3-dimethyl-4-nitropyridine-n-oxide, 4-nitro-2,3-dimethylpyridine n-oxide, 4-nitro-2,3-lutidine n-oxide, 4-nitro-2,3-lutidine-n-oxide, pyridine, 2,3-dimethyl-4-nitro-, 1-oxide
2,3-dimethyl-4-nitro-1-oxidopyridin-1-ium
C7H8N2O3
CFMTVTYBZMKULI-UHFFFAOYSA-N
148223
CC1=C(C=C[N+](=C1C)[O-])[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Documents