Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(n-Undecyloxy)benzoic acid, 98%

$34.01 - $318.64

Chemical Identifiers

Chemical Name or Material 4-(n-Undecyloxy)benzoic acid
CAS 15872-44-3
Molecular Formula C18H28O3
Formula Weight 292.42
MDL Number MFCD00043772
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2472703

  SDS
alfa aesar™
B2472703
1g Each for $34.01

AAB2472706

  SDS
alfa aesar™
B2472706
5g Each for $81.12

AAB2472714

  SDS
alfa aesar™
B2472714
25g Each for $318.64
Description & Specifications

Specifications

Formula Weight 292.42
Chemical Name or Material 4-(n-Undecyloxy)benzoic acid
Assay 98%
Molecular Formula C18H28O3
MDL Number MFCD00043772
CAS 15872-44-3
Boiling Point 96° to 137°C
Quantity 1g