Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(N-Succinimidylmethyl)benzeneboronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 4-(N-Succinimidylmethyl)benzeneboronic acid pinacol ester
Molecular Formula C17H22BNO4
Formula Weight 315.17
MDL Number MFCD16294531
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine-2,5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5190903

 
alfa aesar™
H5190903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5190906

 
alfa aesar™
H5190906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 315.17
Chemical Name or Material 4-(N-Succinimidylmethyl)benzeneboronic acid pinacol ester
Assay 95%
Molecular Formula C17H22BNO4
MDL Number MFCD16294531
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine-2,5-dione
Quantity 1g