Please login to your online account to display your discounted pricing

4-n-Propylphenol, 99%, ACROS Organics™

$29.31 - $29.31

Chemical Identifiers

Chemical Name or Material 4-n-Propylphenol
CAS 645-56-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula CH3CH2CH2C6H4OH
Formula Weight 136.19
MDL Number MFCD00002395
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC131620050

  SDS
thermo fisher scientific
AC131620050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC131620250

  SDS
thermo fisher scientific
AC131620250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC131620010

  SDS
thermo fisher scientific
AC131620010
Glass bottle 1g Each for $29.31
Description & Specifications

Specifications

Formula Weight 136.19
Physical Form Liquid
Assay ≥98.5%
Quantity 5g
Color Clear yellow
Density 0.9830g/mL
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3CH2CH2C6H4OH
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
CAS Min % 98.5
MDL Number MFCD00002395
Molecular Formula C9H12O
Assay Percent Range 99%
CAS 645-56-7
Chemical Name or Material 4-n-Propylphenol
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.