missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(n-Propylcarbamoyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53238MD

 View more versions of this product

Catalog No. AAH53238MD



Specifications

Specifications

4-(n-Propylcarbamoyl)benzeneboronic acid
250mg
C10H14BNO3
4-propylcarbamoyl phenyl boronic acid, 4-n-propylaminocarbonyl phenylboronic acid, 4-propylcarbamoyl phenylboronic acid, 4-propylaminocarbonyl benzeneboronic acid, 4-n-propylcarbamoyl benzeneboronic acid, 4-propylcarbamoyl phenyl boronic acid, n-propyl 4-boronobenzamide, acmc-1c1gn, 4-propylcarbamoyl phenyl boronicacid, 4-n-propylaminocarbonyl phenylboronicacid
B(C1=CC=C(C=C1)C(=O)NCCC)(O)O
207.036
207.04
210°C to 214°C
171922-46-6
MFCD06656015
KWJCBWYHJKWCRD-UHFFFAOYSA-N
[4-(propylcarbamoyl)phenyl]boronic acid
2773569
98%

Chemical Identifiers

171922-46-6
207.036
KWJCBWYHJKWCRD-UHFFFAOYSA-N
2773569
B(C1=CC=C(C=C1)C(=O)NCCC)(O)O
C10H14BNO3
MFCD06656015
4-propylcarbamoyl phenyl boronic acid, 4-n-propylaminocarbonyl phenylboronic acid, 4-propylcarbamoyl phenylboronic acid, 4-propylaminocarbonyl benzeneboronic acid, 4-n-propylcarbamoyl benzeneboronic acid, 4-propylcarbamoyl phenyl boronic acid, n-propyl 4-boronobenzamide, acmc-1c1gn, 4-propylcarbamoyl phenyl boronicacid, 4-n-propylaminocarbonyl phenylboronicacid
[4-(propylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents