Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propylbiphenyl, 98%

$49.86 - $180.81

Chemical Identifiers

Chemical Name or Material 4-n-Propylbiphenyl
CAS 10289-45-9
Molecular Formula C15H16
Formula Weight 196.29
MDL Number MFCD00102114
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1761003

  SDS
alfa aesar™
A1761003
1g Each for $49.86

AAA1761006

  SDS
alfa aesar™
A1761006
5g Each for $180.81
Description & Specifications

Specifications

Formula Weight 196.29
MDL Number MFCD00102114
Quantity 1g
Chemical Name or Material 4-n-Propylbiphenyl
Molecular Formula C15H16
Boiling Point 109° to 111°C (1.5mmHg)
CAS 10289-45-9
Assay 98%
Melting Point 5° to 17°C